Learn More
Heidolph™ Heat-On™ Multi-Well Holder Insert
GREENER_CHOICE
Insert for Heidolph Heat-On Multi-Well Holders when used with Hei-PLATE magnetic stirring hotplates.
Supplier:  Heidolph™ 015882110
 View more versions of this product
Catalog No. 05404167
Only null left
Description
Description
  • Inserts are available for 10 mL, 25 mL, 50 mL, 100 mL and 150 mL flasks
  • Flask inserts also feature cut-away sides for use with two or three neck flasks
  • Multi-tube inserts are available for 16 mm, 20 mm, and 24 mm tubes
  • Available with fluoropolymer coating or anodized finish
Specifications
Chemical Identifiers
Heat-On Insert
Specifications
Multi-Well Holder, Hei Series Hotplate
8 x 16 mm Tubes
Heat-On Insert
Fluoropolymer
Product Suggestions
Product Suggestions