Learn More
Synthware™ Heavy Wall Cylindrical Pressure Vessels
GREENER_CHOICE
Cylindrical vessels feature a protective coating with a working temperature ranging from -40°C to 80°C.
$220.32 - $283.36
Specifications
Material Glass
Includes Teflon™ Bushing, EP™ O-Ring
Max. Pressure 88 psi
Shape Cylindrical
Surface Coating Protective
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
31502243
GREENER_CHOICE
View Documents Promotion Details
Synthware™
P160001D
15 mL
Each for $220.32
Only null left
31502244
GREENER_CHOICE
View Documents Promotion Details
Synthware™
P160002D
35 mL
Each for $222.57
Only null left
31502245
GREENER_CHOICE
View Documents Promotion Details
Synthware™
P160003D
38 mL
Each for $225.50
Only null left
31502246
GREENER_CHOICE
View Documents Promotion Details
Synthware™
P160004D
48 mL
Each for $269.54
Only null left
31502247
GREENER_CHOICE
View Documents Promotion Details
Synthware™
P160005D
100 mL
Each for $271.82
Only null left
31502248
GREENER_CHOICE
View Documents Promotion Details
Synthware™
P160006D
120 mL
Each for $283.36
Only null left
Description
Description
  • Protective coating has a working temperature ranging from -40°C to 80°C
  • Coating may yellow over time, but will not affect the vessel's integrity
  • Complete apparatus is supplied with a vessel, Teflon™ bushing, and an EP™ O-ring
  • Always work behind a safety shield since there is a possibility that glass vessels can break under pressure
Specifications
Specifications
Glass
88 psi
Protective
Teflon™ Bushing, EP™ O-Ring
Cylindrical
#15