missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Heavy Duty Mounting Clips

Catalog No. 19111923
Change view
Click to view available options
Material:
Metal, Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19111923 Metal, Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19111923 Supplier Brady™ Supplier No. 42335

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with HPHV pipe markers.

Specifications

Material Metal, Plastic
Quantity 1 Ea.
Product Type Clip
Color Black