Learn More
Chemglass Inc 500mL Glas-Col Heating Mantle, Series O, Hemispherical, 270 Watts
Numéro de catalogue. CG1000007
Encompass
500mL Glas-Col Heating Mantle, Series O, Hemispherical, 270 Watts. The Glas-Col Series O mantle is a hemispherical style, meaning it covers only the bottom half of the flask. This allows full view of contents. The mantle accommodates single and multi-flasks. It should be used with the Series O supports. All mantles are 115 Volts and are supplied with a separable 4 foot, 2 wire cord and locking connector. For Series O supports see CG-10010 or CG-10012.
Numéro de catalogue. CG1000007 Fournisseur Chemglass Inc Code fournisseur CG1000007
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
500mL Glas-Col Heating Mantle, Series O, Hemispherical, 270 Watts. The Glas-Col Series O mantle is a hemispherical style, meaning it covers only the bottom half of the flask. This allows full view of contents. The mantle accommodates single and multi-flasks. It should be used with the Series O supports. All mantles are 115 Volts and are supplied with a separable 4 foot, 2 wire cord and locking connector. For Series O supports see CG-10010 or CG-10012.