Learn More
StatLab HCS Gram ± Control Slide
Numéro de catalogue. 22024780
Modifier l'affichage
Click to view available options
Quantité:
25/pk.
Numéro de catalogue. Quantité
22024780 25/pk.
1 options
Numéro de catalogue. 22024780 Fournisseur StatLab Code fournisseur CS040PS25
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Adhesive control slides for evaluating Gram stain quality and performance in histology laboratories.
  • Contains formalin-fixed, paraffin-embedded tissue sections with known positive tissue elements
  • Designed to evaluate Gram stain quality and staining performance in histology laboratories
  • Manufactured using adhesion white glass slides with 90° corners
  • Includes a specific staining protocol with each package for consistent use
  • Provides reliable quality control for histology staining procedures
Spécifications
Type de produit Control Slide
Quantité 25/pk.