missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ N, 100-120 mesh

Catalog No. 111004194
Change view
Click to view available options
Quantity:
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004194 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004194 Supplier MilliporeSigma™ Supelco™ Supplier No. 10296

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HayeSep™ N was used as a adsorbent for analytical preconcentration of methane which contributes majorly to the greenhouse effect. It was found to have good adsorption capacity, highest overall selectivity and highest enthalpy for methane adsorption as compared to other adsorbents.

HayeSep™ porous polymers, considered second generation materials, are consistent batch-to-batch, with minimal shrinkage and monomer bleed.

Specifications

Particle Size 100/120 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) ˜405 m2/g
Product Line HayeSep
For Use With (Application) GC
Density ˜0.36 g/mL (free fall density)

HayeSep is a registered trademark of Hayes Separation Inc.