Learn More
Haws™ Hose Units
Catalog No. 17162C
Change view
Click to view available options
Mounting:
Wall
Catalog No. Mounting
17162C Wall
1 options
Catalog No. 17162C Supplier Haws™ Supplier No. 8901B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Wall mounting bracket.
Assists in irrigation process
  • Dust cover allows for spray head to stay protected from debris when not in use
  • Requires minimal maintenance and is equipped with test card to record weekly testing
  • 8 ft. (243.8cm) swivel hose provides extended coverage area to further optimiz irrigation process
  • Drench head can be easily activated with integral squeeze lever
  • Conveniently mounted using wall clips
Compliance
Certified by CSA to meet the ANSI Z358.1 Standard for Emergency Eyewash and Shower Equipment
Specifications
Certifications/Compliance ANSI Z358.1, CSA
For Use With (Application) Maximum Coverage area for the Eye, Face, or Body
Includes Green ABS Plastic head, Chrome Plated Brass stay open squeeze lever valve, Green Rubber hose, wall mounting bracket, universal sign
Length (English) 8 ft.
Length (Metric) 2.43 m
Mounting Wall
Product Type Hose Unit