missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haws™ Cleansing Stick
Description
Model 9084 is an immersible Cleansing Stick that gives continual protection against pathogenic bacteria for water storage systems. It balances the PH of the water preventing the build-up of bio-film, scale and corrosion for up to 3 years with proper usage. This non-chemical disinfecting module is designed to be immersed in a water tank to provide constant protection from bio-film and hazardous levels of pathogenic bacteria, including Legionella. The Cleansing Stick will treat up to 528 gallons (2000 liters). Dimensions: 32 in. (81.3 cm) long x 3 in. (7.5 cm) in diameter.
Warranty
1 year from ship date (Limited)
Specifications
Specifications
| Color | Yellow |
| For Use With (Equipment) | 8740, Emergency Overhead Tank Shower |
| Height (English) | 3 in. |
| Height (Metric) | 7.62 cm |
| Product Type | Cleansing Stick |
| Weight (English) | 7 lb. |
| Weight (Metric) | 3.17515 kg |