missing translation for 'onlineSavingsMsg'
Learn More

Hastelloy C∣r gauze, 100 mesh woven from 0.102mm (0.004in) dia wire

Catalog No. AA40940GT
Change view
Click to view available options
Quantity:
150 x 150 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40940GT 150 x 150 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40940GT Supplier Thermo Scientific Chemicals Supplier No. 040940GT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Hastelloy C™ gauze
Name Note Woven from 0.102mm (0.004 in.) dia. wire
Quantity 150 x 150 mm
Mesh Size 100 Mesh
Form Wire Cloth
MDL Number MFCD00801065

RUO – Research Use Only