missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAS1 Monoclonal Antibody (5C9D6)

Description
HAS1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Spécifications
Spécifications
| Antigen | HAS1 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 5C9D6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | Has1 |
| Gene Accession No. | Q61647, Q92839 |
| Gene Alias | HA synthase 1; HAS; Has1; huHAS1; Hyaluronan synthase 1; hyaluronan synthase1; hyaluronate synthase 1; Hyaluronic acid synthase 1 |
| Afficher plus de résultats |