Learn More
Hanna™ Instruments Zero Oxygen Solution
Numéro de catalogue. 13299152
Encompass
Change view
Click to view available options
For Use With (Equipment):
Polarographic DO Sensors, Galvanic DO Sensor
Numéro de catalogue. For Use With (Equipment)
13299152 Polarographic DO Sensors, Galvanic DO Sensor
1 options
Numéro de catalogue. 13299152 Fournisseur Hanna Instruments CDA Inc Code fournisseur HI 7040L
Il en reste null
High quality, bicomponent zero oxygen solutions made from reagent grade chemicals that are contained in hassle-free, easy to mix chemical component bottles.
  • High quality, bicomponent zero oxygen solution made from reagent grade chemicals
  • Prepared with reagent grade chemicals
  • Hassle-free, easy to mix chemical component bottles
  • Air-tight seal to ensure the quality of the solution
  • Bicomponent zero oxygen solution designed for calibrating both polarographic and galvanic dissolved oxygen sensors
Spécifications
For Use With (Equipment) Polarographic DO Sensors, Galvanic DO Sensor