missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Hand Lens, Plastic Frame

Durable design for repeated use.

Supplier:  United Scientific™ MFP090

 Afficher plus de versions de ce produit

Numéro de catalogue. S15971


Il en reste null

Description

Description

  • Glass lens, molded into high-quality plastic frame.
  • Available in various diameters
  • Provides 2.5x magnification
Spécifications

Identifiants chimiques

2.5x
Glass, Plastic
Magnifier

Spécifications

Magnifier
90 mm
2.5x
Glass, Plastic
3.5 in.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

Warning: Cancer and reproductive harm