Learn More
Hamilton™ SampleLock Syringe, Sold by MilliporeSigma™ Supelco™
Connects to either male or female luer fittings by using an adapter
Supplier:  Merck Emd Millipore 26282
 View more versions of this product
Catalog No. 111010208
Only null left
Description
Description
  • Positive rear plunger stop eliminates sample loss and plunger blowout at high pressures (not applicable to 1705 and 1710 models)
  • Connects to either male or female luer fittings by using an adapter
  • Male and female luer and male luer lock adapters thread onto the SampleLock Valve
  • Syringe has the possibility to be compatible with many connectors and fittings
  • All adapters have a Kel-F (CTFE) fluid path, with a port diameter of 0.039 in.
  • Internal standards can be added through the valve port into the sample contained in the syringe
  • Twist valve locks the sample in the syringe
Specifications
Chemical Identifiers
Not Autoclavable
0.0283 in.
2 in.
50°F
10°C
Hamilton
Bevel
Borosilicate Glass (Barrel), PTFE (Plunger), Stainless Steel (Tip)
0.72 mm
Gastight™ Syringe
2
1 Ea.
51 mm
Sample Lock
Specifications
Not Autoclavable
22s G
0.15 mm
0.72 mm
Gastight™ Syringe
239°F
50°F
2
1 Ea.
Hamilton
Bevel
Borosilicate Glass (Barrel), PTFE (Plunger), Stainless Steel (Tip)
0.0059 in.
0.0283 in.
2 in.
500 psig (34.5 bar)
115°C
1725 SL
10°C
250 μL
51 mm
Sample Lock
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Hamilton is a registered trademark of Hamilton Co.