Learn More
Hamilton™ Preflanged PTFE Tubing Assemblies
Catalog No. 361012603
Change view
Click to view available options
Product Type:
Tubing Assembly; 1/4-28 NT (SR)
Tubing Assembly; 1M6X1 FTG TPRD TIP
Overall Length (Metric):
650 mm
762 mm
Gauge:
12 ga
18 ga
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Overall Length (Metric) Gauge
361012603 Tubing Assembly; 1M6X1 FTG TPRD TIP 650 mm 18 ga
14819163 Tubing Assembly; 1/4-28 NT (SR) 762 mm 12 ga
14819164 Tubing Assembly; 1/4-28 NT (SR) 762 mm 18 ga
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 361012603 Supplier Hamilton™ Supplier No. 88966
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Pre-made flanged tubing assemblies made with PTFE tubing and Kel-F threaded fittings. The flanged tubing assemblies provide the perfect seal with 1/4-28 flat bottom valve ports.
Specifications
Material PTFE
Overall Length (Metric) 650 mm
Includes 0.25 to 28 flat bottom valve ports
Gauge 18 ga
Product Type Tubing Assembly; 1M6X1 FTG TPRD TIP