missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Hamilton™ Needles for LT and TLL Syringes, KEL-F hub (KF)
GREENER_CHOICE

$107.58 - $112.68

Specifications

Material Stainless Steel (Tip)
Description Has KEL-F hub (KF)
Includes KEL-F hub (KF)
Length (English) Needle 2 in.
Quantity 6/Pk.
View More Specs

Products 3
Catalog Number Mfr. No. Needle Gauge Needle Point Style Product Type Price Quantity  
Catalog Number Mfr. No. Needle Gauge Needle Point Style Product Type Price Quantity  
111011339
View Documents
MilliporeSigma™ Supelco™
21747
22 G 2 Bevel Tip
Each for $109.19
Only null left
 
111011338
View Documents
MilliporeSigma™ Supelco™
21744
22 G 3 Blunt Tip
Each for $107.58
Only null left
 
111011269
View Documents
MilliporeSigma™ Supelco™
Z2889771PAK
22s G 3 Blunt Tip
Each for $112.68
Only null left
 
Description

Description

Specifications

Specifications

Stainless Steel (Tip)
KEL-F hub (KF)
6/Pk.
Hamilton LT and TLL Syringes
Has KEL-F hub (KF)
2 in.
51 mm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Hamilton is a registered trademark of Hamilton Co.