missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Hamilton™ Needles for LT and TLL Syringes, KEL-F hub (KF)
Supplier: MilliporeSigma™ Supelco™ Z2889771PAK
Specifications
| Stainless Steel (Tip) | |
| 22s G | |
| 2 in. | |
| 6/Pk. | |
| 3 |
| Has KEL-F hub (KF) | |
| KEL-F hub (KF) | |
| Blunt Tip | |
| 51 mm | |
| Hamilton LT and TLL Syringes |
Hamilton is a registered trademark of Hamilton Co.