missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Hamilton™ Needles for LT and TLL Syringes, KEL-F hub (KF)

Catalog No. 111011339
Change view
Click to view available options
Needle Gauge:
22s G
22 G
Needle Point Style:
2
3
Product Type:
Bevel Tip
Blunt Tip
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Needle Point Style Product Type
111011339 22 G 2 Bevel Tip
111011338 22 G 3 Blunt Tip
111011269 22s G 3 Blunt Tip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111011339 Supplier MilliporeSigma™ Supelco™ Supplier No. 21747

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Material Stainless Steel (Tip)
Description Has KEL-F hub (KF)
Needle Gauge 22 G
Includes KEL-F hub (KF)
Length (English) Needle 2 in.
Product Type Bevel Tip
Quantity 6/Pk.
Length (Metric) Needle 51 mm
Needle Point Style 2
For Use With (Equipment) Hamilton LT and TLL Syringes

Hamilton is a registered trademark of Hamilton Co.