Learn More
Hamilton™ HPLC Syringes, Sold by MilliporeSigma™ Supelco™
May be used for extraction and injection of sample material
Supplier:  Merck Emd Millipore 20889
 View more versions of this product
Catalog No. 111011071
Only null left
Description
Description
Specifications
Specifications
Autoclavable
22 G
0.41 mm
0.72 mm
Gastight™ Syringe
239°F
50°F
3
1 Ea.
Hamilton
Blunt
Removable
Borosilicate Glass (Barrel), PTFE (Plunger), Stainless Steel (Tip)
0.0161 in.
0.0283 in.
2 in.
500 psig (34.5 bar)
115°C
1725 RN
10°C
250 μL
51 mm
Rheodyne, SSI, and Valco VISF-2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Hamilton is a registered trademark of Hamilton Co.