Learn More
Hamilton™ Gastight™ Syringe, 50 μL, Sold by MilliporeSigma™ Supelco™
Gas-tight design minimizes gas exchange during GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed.
Supplier:  Merck Emd Millipore SGMA26202
 View more versions of this product
Catalog No. 111011263
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Autoclavable
1000 psig (68.9 bar)
115°C
1705 TLL
1 Ea.
Hamilton
Borosilicate Glass (Barrel), PTFE (Plunger)
239°F
50°F
10°C
50 μL
PTFE Luer Lock (TLL) without Slots
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
GASTIGHT is a registered trademark of Hamilton Co.