Learn More
Hamilton™ Gastight™ Syringe, 250 μL, Sold by MilliporeSigma™ Supelco™
Gas-tight design minimizes gas exchange during GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed.
Supplier: Merck Emd Millipore 20705
Specifications
| Borosilicate Glass (Barrel), PTFE (Plunger), Stainless Steel (Tip) | |
| 22s G | |
| 0.15 mm | |
| 0.72 mm | |
| Gastight™ Syringe | |
| 122°F | |
| 50°F | |
| 5 | |
| 1 Ea. | |
| Hamilton | |
| Cone with Side Port |
| Luer Tip Cemented Needle (LTN), Cone Tip/Side Port | |
| 0.0059 in. | |
| 0.0283 in. | |
| 2 in. | |
| 500 psig (34.5 bar) | |
| 50°C | |
| 1725 LTN | |
| 10°C | |
| 250 μL | |
| 51 mm | |
| Luer Tip, Cemented |
GASTIGHT is a registered trademark of Hamilton Co.