Learn More
Hamilton™ 1700 Series Gastight™ Syringes: LT Termination
Gastight version of the original 700 series syringes, ideal for dispensing volumes from 1 up to 500μL
$85.94 - $111.02
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass (Barrel), PTFE (Tip)
For Use With (Application) Ideal for dispensing both liquids and gases, useful for samples with dissolved or suspended molecules
Product Type Gastight
Max. Temperature (English) 239°F
View More Specs
Products 6
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Inner Diameter (Metric) Outer Max. Pressure Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Diameter (Metric) Inner Diameter (Metric) Outer Max. Pressure Price Quantity  
13-684-104
View Documents
Hamilton™
HAM81201
500 μL 3.26 mm 7.75 mm 700 psig
Each for $111.02
Only null left
14-815-2
View Documents
Hamilton™
HAM80001
10 μL 0.46 mm 6.6 mm 1000 psig
Each for $85.94
Only null left
14-815-30
View Documents
Hamilton™
HAM80201
25 μL 0.72 mm 7.75 mm 1000 psig
Each for $93.11
Only null left
14-815-75
View Documents
Hamilton™
HAM81001
100 μL 1.46 mm 7.75 mm 1000 psig
Each for $93.11
Only null left
14-815-92
View Documents
Hamilton™
HAM81101
250 μL 2.3 mm 7.75 mm 500 psig
Each for $98.47
Only null left
14-815-50
View Documents
Hamilton™
HAM80901
50 μL 1.03 mm 7.75 mm 1000 psig
Each for $93.11
Only null left
Description
Description
Syringe series offers a variety of different terminations and needle options.
Luer Tip (LT)- The needles are removable and fit over a ground glass hub which is tapered in the shape of a male luer. The LT termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
Needle Point Styles (Point styles 4 and AS are available on request)
  • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 34-10
  • Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 34-10
  • Point Style 4 - Standard 12° beveled needle point is recommended for life science applications. Available gauges: 34-10. Special needle angles such as 0° to 60° available upon request.
  • Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-10
  • Point Style AS - Special conical style needle point used on autosampler syringes the non-coring needle point is recommended for septum penetration. Available gauges: 26-10
Specifications
Specifications
Autoclavable
Ideal for dispensing both liquids and gases, useful for samples with dissolved or suspended molecules
239°F
50°F
10°C
Luer Tip
Borosilicate Glass (Barrel), PTFE (Tip)
Gastight
115°C
No Needle Included
Ea.