missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ 1000 Series Syringe, Without Needle, Sold by MilliporeSigma™ Supelco™
Gas-tight design minimizes the gas exchange in GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed. Tip is threaded in a luer lock syringe which allows the needle to be tightly screwed to the tip.
Supplier: Merck Emd Millipore SGMA20997
Chemical Identifiers
| Gastight™ Syringe | |
| 1 Ea. | |
| Hamilton |
Specifications
| Autoclavable | |
| 0.181 in. | |
| 4.61 mm | |
| Gastight™ Syringe | |
| 239°F | |
| 50°F | |
| 10°C | |
| 1 mL | |
| Hamilton 1001 TLL |
| Borosilicate Glass (Barrel), PTFE (Plunger) | |
| 0.355 in. | |
| 9.02 mm | |
| 200 psig (13.8 bar) | |
| 115°C | |
| 1001 TLL | |
| 1 Ea. | |
| Hamilton | |
| Metal (N) or Kel-F |
GASTIGHT is a registered trademark of Hamilton Co.