Learn More
Hamilton™ 1000 Series Syringe, Without Needle, Sold by MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Type | Gastight™ Syringe |
| Model | 1025 TLL |
| Quantity | 1 Ea. |
| Volume (Metric) | 25 mL |
| Product Line | Hamilton |
| For Use With (Equipment) | Hamilton 1025 TLL |
| Connection Type | Kel-F Luer Lock with Slots |
GASTIGHT is a registered trademark of Hamilton Co.