Learn More
Hamilton™ 1000 Series Syringe, Without Needle, Sold by MilliporeSigma™ Supelco™
Gas-tight design minimizes the gas exchange in GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed. Tip is threaded in a luer lock syringe which allows the needle to be tightly screwed to the tip.
Supplier:  Merck Emd Millipore 20722
 View more versions of this product
Catalog No. 111011026
Only null left
Description
Description
Specifications
Chemical Identifiers
Gastight™ Syringe
1 Ea.
Hamilton
Specifications
Autoclavable
0.406 in.
10.3 mm
Gastight™ Syringe
239°F
50°F
1 Ea.
Hamilton
Borosilicate Glass (Barrel), PTFE (Plunger)
0.530 in.
13.5 mm
200 psig (13.8 bar)
115°C
10°C
5 mL
Luer Tip, Kel-F Hub
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
GASTIGHT is a registered trademark of Hamilton Co.