Learn More
Hamilton™ 1000 Series Syringe, Without Needle, Sold by MilliporeSigma™ Supelco™
Gas-tight design minimizes the gas exchange in GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed. Tip is threaded in a luer lock syringe which allows the needle to be tightly screwed to the tip.
Supplier: Merck Emd Millipore 20683
Chemical Identifiers
| Gastight™ Syringe | |
| 1 Ea. | |
| Hamilton |
Specifications
| Not Autoclavable | |
| 0.907 in. | |
| 23 mm | |
| Gastight™ Syringe | |
| 176°F | |
| 50°F | |
| 10°C | |
| 25 mL | |
| Metal (N) or Kel-F |
| Borosilicate Glass (Barrel), PTFE (Plunger) | |
| 1.07 in. | |
| 27.1 mm | |
| 100 psig (6.9 bar) | |
| 80°C | |
| 1025 TLL | |
| 1 Ea. | |
| Hamilton |
GASTIGHT is a registered trademark of Hamilton Co.