Learn More
Hamilton™ 1000 Series Syringe, Without Needle, Sold by MilliporeSigma™ Supelco™
Gas-tight design minimizes the gas exchange in GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed. Tip is threaded in a luer lock syringe which allows the needle to be tightly screwed to the tip.
Supplier:  Merck Emd Millipore 20707
 View more versions of this product
Catalog No. 111009485
Only null left
Description
Description
Specifications
Chemical Identifiers
Gastight™ Syringe
1 Ea.
Hamilton
Specifications
Not Autoclavable
1.28 in.
32.6 mm
Gastight™ Syringe
176°F
50°F
10°C
50 mL
Metal (N) or Kel-F
Borosilicate Glass (Barrel), PTFE (Plunger)
1.45 in.
36.9 mm
100 psig (6.9 bar)
80°C
1050 TLL
1 Ea.
Hamilton
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
GASTIGHT is a registered trademark of Hamilton Co.