Learn More
Hamilton™ 1000 Series Syringe, Without Needle, Sold by MilliporeSigma™ Supelco™
Gas-tight design minimizes the gas exchange in GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed. Tip is threaded in a luer lock syringe which allows the needle to be tightly screwed to the tip.
Supplier:  Merck Emd Millipore 26213
 View more versions of this product
Catalog No. 111009346
Only null left
Description
Description
Specifications
Chemical Identifiers
Gastight™ Syringe
1 Ea.
Hamilton
Specifications
Gastight™ Syringe
1 Ea.
Hamilton
Kel-F Luer Lock with Slots
1050 TLL
50 mL
Hamilton 1050 TLL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
GASTIGHT is a registered trademark of Hamilton Co.