missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ 1000 Series Syringe, Without Needle, Sold by MilliporeSigma™ Supelco™
Gas-tight design minimizes the gas exchange in GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed. Tip is threaded in a luer lock syringe which allows the needle to be tightly screwed to the tip.
Supplier: Merck Emd Millipore 21967
Chemical Identifiers
| Gastight™ Syringe | |
| 1 Ea. | |
| Hamilton |
Specifications
| Not Autoclavable | |
| 1.28 in. | |
| 32.6 mm | |
| Gastight™ Syringe | |
| 176°F | |
| 50°F | |
| 10°C | |
| 100 mL | |
| Hamilton 1100 TLL |
| Borosilicate Glass (Barrel), PTFE (Plunger) | |
| 1.45 in. | |
| 36.9 mm | |
| 100 psig (6.9 bar) | |
| 80°C | |
| 1100 TLL | |
| 1 Ea. | |
| Hamilton | |
| Metal (N) or Kel-F |
GASTIGHT is a registered trademark of Hamilton Co.