missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Halt™ Protease Inhibitor Cocktail (100X)

Catalog No. PI78430
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
24 x 100 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI78430 24 x 100 μL
PI78429 5 mL
PI78438 10 mL
PI87786 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI78430 Supplier Thermo Scientific™ Supplier No. 78430

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibit proteolytic degradation during cell lysis and protein extraction with these ready-to-use concentrated stock solutions of protease inhibitors.

Thermo Scientific™ Halt Protease Inhibitor Cocktails are ready-to-use concentrated stock solutions of protease inhibitors for addition to samples to prevent proteolytic degradation during cell lysis and protein extraction procedures.

Halt Protease Inhibitor Cocktails are 100X solutions containing optimized concentrations of six broad-spectrum protease inhibitors stabilized in high-quality dimethylsulfoxide (DMSO). The cocktails effectively inhibit serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The identity and concentration of each component is fully disclosed, leaving no uncertainties about variables and compatibility with experiments. Regular and EDTA-free formulations provide for inhibition of metalloproteases and compatibility with 2D electrophoresis, respectively. The stable, concentrated liquid cocktails are easy to use, and the several different package sizes make Halt Protease Inhibitor Cocktails exceptionally convenient for either small-scale or large-scale uses.

Highlights:

  • Better than tablets – the refrigerator-stable, 100X liquid format is more effective and easier to use than individual inhibitors or other formats; just pipette exactly the amount you need
  • No proprietary ingredients – fully disclosed formulation contains AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A in DMSO
  • Multiple package sizes – choose among several package sizes, including single-use microtubes that keep protease inhibitors fresh and reduce the risk of reagent contamination
  • Two popular formulations – regular formulation includes a separate vial of EDTA solution for optional metalloprotease inhibition; EDTA-free formulation ensures compatibility with isoelectric focusing
  • Lysis buffer compatible – use with Thermo Scientific Pierce™ Cell Lysis Buffers or nearly any other commercial or homemade detergent-based lysis reagent

Includes:

Protease inhibitor cocktails with EDTA include separate 0.5M EDTA solution.

Specifications

Product Type Protease Inhibitor Cocktail
Content And Storage Upon receipt store at 4°C
Form Liquid
Product Line Halt
Buffer Lysis Buffer
Reagent Type Protease Inhibitor
Sufficient For 240 mL Sample
Inhibitors Protease Inhibitor
Quantity 24 x 100 μL
What are the compositions of the Halt Protease Inhibitor Cocktail, EDTA-Free (100X) (Cat. No. 87785) and Halt Protease Inhibitor Cocktail (100X) (Cat. No. 87786)?

Both formulations are the same and have the following composition:

  • AEBSF – 100mM
  • Aprotinin – 80µM
  • Bestatin – 5mM
  • E64 – 1.5mM
  • Leupeptin – 2mM
  • Pepstatin A – 1mM
What is the shelf life of Halt Protease Inhibitor Cocktail (100X)?

When stored as recommended at 4 degrees C, Halt Protease Inhibitor Cocktail (100X) is guaranteed stable for 1 year from the date of receipt.

Can I use just part of the Pierce Protease Inhibitor XL Capsules, EDTA-Free and save part of the capsule for later use?

We do not recommend “splitting” the capsule into smaller portions. Researchers using less than 500 mL should choose a different format (liquid or tablet) of protease inhibitors.

What formats of protease inhibitors do you offer?

We offer standalone protease inhibitors (AEBSF, Aprotinin, PMSF, EDTA, etc.), as well as cocktails or combinations of protease inhibitors in liquid, tablet, and larger capsule formats. These cocktails of protease inhibitors can also be purchased in combination with a cocktail of phosphatase inhibitors, in both liquid and tablet form, for even greater convenience.

What protease inhibtors are present in the Halt Protease Inhibitor Single-Use Cocktail?

The inhibitors included in the Halt Protease Inhibitor Single-Use Cocktail (Cat. No. 78430) are: AEBSF, Aprotinin, Bestatin, E64, Leupeptin, Pepstatin A, and EDTA.

Does the Halt Protease Inhibitor Single-Use Cocktail contain phosphatase inhibitors?

No, the Halt Protease Inhibitor Single-Use Cocktail is a broad-spectrum protease inhibitor that does not contain phosphatase inhibitors.


For Research Use Only. Not for use in diagnostic procedures.