missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Halt™ Protease Inhibitor Cocktail, EDTA-Free (100X)

Catalog No. p-9056012
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
24 x 100 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI78425 24 x 100 μL
PI78437 5 mL
PI78439 10 mL
PI87785 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI78425 Supplier Thermo Scientific™ Supplier No. 78425

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibit proteolytic degradation during cell lysis and protein extraction with these ready-to-use concentrated stock solutions of protease inhibitors.

Thermo Scientific Halt Protease Inhibitor Cocktail, EDTA-free (100X) are ready-to-use concentrated stock solutions of protease inhibitors for addition to samples to prevent proteolytic degradation during cell lysis and protein extraction.

Features of Halt Protease Inhibitor Cocktail, EDTA-free (100X):

  • Convenient—the refrigerator-stable, 100X liquid format is more effective and easier to use than individual inhibitors or other formats; just pipette exactly the amount you need
  • No proprietary ingredients—fully disclosed formulation contains optimized concentrations of six broad-spectrum protease inhibitors AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A stabilized in high-quality dimethylsulfoxide (DMSO)
  • Multiple package sizes—choose among several package sizes, including single-use microtubes that keep protease inhibitors fresh and reduce the risk of reagent contamination
  • EDTA-free formulations—ensures compatibility with isoelectric focusing
  • Lysis buffer compatible—use with Thermo Scientific Pierce Cell Lysis Buffers or nearly any other commercial or homemade detergent-based lysis reagent. Add 10 μL of concentrated cocktail per 1 mL of lysis buffer to ensure complete protection of the resulting protein extract.

Applications:

  • Protection of intact, active cellular proteins from degradation by endogenous proteases
  • Inhibition of protease activity from lysates produced by a variety of methods including Thermo Scientific Pierce Cell Lysis Reagents, other commercially formulated cell or tissue lysis products, sonication, French press, blender homogenization or repetitive freeze/thaw cycling
  • Screening of extracts for proteolytic activity
  • The study of proteolysis in the regulation of cellular processes

Proteases make up a large category of enzymes, including endopeptidases and exopeptidases, which catalyze the hydrolytic breakdown of proteins into peptides or amino acids in a wide variety of cell and tissue types. Proteases are regulated and compartmentalized in living cells, but cell lysis releases and mixes them with other extracted proteins. Their activity can thus lead to the loss of a large number of valuable proteins, adversely affecting downstream applications. For this reason, it is common practice to spike cell lysis buffers or cell extracts with a cocktails of compounds known to inactivate or inhibit proteases. Halt Protease Inhibitor Cocktails effectively inhibit serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. Regular and EDTA-free formulations provide for inhibition of metalloproteases and compatibility with 2D electrophoresis, respectively.

Related Products

  • Halt™ Protease Inhibitor Single-Use Cocktail (100X) (Cat. No. 78430)

Specifications

Product Type Protease Inhibitor Cocktail
Content And Storage Upon receipt store at 4°C
Form Liquid
Product Line Halt
Buffer Lysis Buffer
Reagent Type Protease Inhibitor
Inhibitors Protease Inhibitor
Quantity 24 x 100 μL
What are the compositions of the Halt Protease Inhibitor Cocktail, EDTA-Free (100X) (Cat. No. 87785) and Halt Protease Inhibitor Cocktail (100X) (Cat. No. 87786)?

Both formulations are the same and have the following composition:

  • AEBSF – 100mM
  • Aprotinin – 80µM
  • Bestatin – 5mM
  • E64 – 1.5mM
  • Leupeptin – 2mM
  • Pepstatin A – 1mM
What is the shelf life of Halt Protease Inhibitor Cocktail, EDTA-free (100X) (Cat. No. 78425)?

The Halt Protease Inhibitor Cocktail, EDTA-free (100X) (Cat. No. 78425) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale for more details.

Can I use just part of the Pierce Protease Inhibitor XL Capsules, EDTA-Free and save part of the capsule for later use?

We do not recommend “splitting” the capsule into smaller portions. Researchers using less than 500 mL should choose a different format (liquid or tablet) of protease inhibitors.

What formats of protease inhibitors do you offer?

We offer standalone protease inhibitors (AEBSF, Aprotinin, PMSF, EDTA, etc.), as well as cocktails or combinations of protease inhibitors in liquid, tablet, and larger capsule formats. These cocktails of protease inhibitors can also be purchased in combination with a cocktail of phosphatase inhibitors, in both liquid and tablet form, for even greater convenience.

For Research Use Only. Not for use in diagnostic procedures.