missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Halt™ Phosphatase Inhibitor Single-Use Cocktail
Stabilize serine, threonine and tyrosine phosphorylation of proteins in cells and tissues with this cocktail of phosphatase inhibitors.
$532.06 - $2412.35
Specifications
| Product Type | Phosphatase Inhibitor Cocktail |
|---|---|
| Content And Storage | Upon receipt store product at 4°C. Do not freeze. |
| Form | Liquid |
| Product Line | Halt |
| Reagent Type | Phosphatase Inhibitor |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
PI78420
|
Thermo Scientific™
78420 |
1 mL |
Each for $532.06
|
|
|||||
|
PI78428
|
Thermo Scientific™
78428 |
24 x 100 μL |
Each for $882.44
|
|
|||||
|
PI78426
|
Thermo Scientific™
78426 |
5 x 1 mL |
Each for $1,752.97
|
|
|||||
|
PI78427
|
Thermo Scientific™
78427 |
10 mL |
Each for $2,412.35
|
|
|||||
Description
Preserve phosphorylation of proteins in cells and tissues with Thermo Scientific™ Halt Phosphatase Inhibitor Cocktail.
- Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from serine/threonine phosphatases, as well as protein tyrosine phosphatases (PTPs) during cell lysis
- The cocktail contains a mixture of four inhibitors of broad specificity including sodium fluoride, sodium orthovanadate, sodium pyrophosphate, and beta-glycerophosphate
- Conveniently packaged in a 100X format, the cocktail is stable at 4°C and is suitable for use with any cell or tissue extract
- The cocktail preserves protein phosphorylation in immunoprecipitation and kinase assays
- Phosphorylation and dephosphorylation comprise a molecular on/off switch that regulates many key biological pathways within the cell including signal transduction, cell division, and apoptosis
- To effectively study phosphorylation activation states, one must inactivate phosphatases to prevent their uncontrolled activity upon cell lysis
Additional Features
- Devoid of protease inhibitors – To protect against both proteases and phosphatases, use Halt Protease and Phosphatase Inhibitor Cocktail (78420)
- Dialyzable – Either dialyze or desalt sample to effectively remove inhibitors from cell extracts before performing procedures that are discovered to be incompatible with cocktail components
- Supplied as a 100X concentrate – effective when used at a final concentration of 1X; however, samples that contain particularly high levels of phosphatase activity may require a final concentration of 2-3X
- Compatibility – compatible with most detergent-based cell lysis reagents, including all Thermo Scientific Pierce Cell Lysis Solutions
- All-in-one format – inhibits both serine-threonine phosphatases and protein tyrosine phosphatases
- Compatible with popular protein assays – works with standard protein assays, including BCA and coomassie (Bradford) assays
- Convenient storage – refrigerator storage (2 to 8°C) saves valuable space in the freezer and allows for immediate use
Specifications
| Phosphatase Inhibitor Cocktail | |
| Liquid | |
| Phosphatase Inhibitor |
| Upon receipt store product at 4°C. Do not freeze. | |
| Halt | |
| Phosphatase Inhibitor |
For Research Use Only. Not for use in diagnostic procedures.