missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Halt™ Phosphatase Inhibitor Single-Use Cocktail

Catalog No. PI78420
Change view
Click to view available options
Quantity:
1 mL
10 mL
24 x 100 μL
5 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI78420 1 mL
PI78428 24 x 100 μL
PI78426 5 x 1 mL
PI78427 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI78420 Supplier Thermo Scientific™ Supplier No. 78420

Stabilize serine, threonine and tyrosine phosphorylation of proteins in cells and tissues with this cocktail of phosphatase inhibitors.

Thermo Scientific Halt Phosphatase Inhibitor Single Use Cocktail preserves the phosphorylation state of proteins during and after cell lysis or tissue protein extraction.

Features of Halt Phosphatase Inhibitor Single Use Cocktail:

  • Devoid of protease inhibitors—To protect against both proteases and phosphatases, use Halt Protease and Phosphatase Inhibitor Cocktail (Cat. No. 78420)
  • Dialyzable— Either dialyze or desalt sample to effectively remove inhibitors from cell extracts before performing procedures that are discovered to be incompatible with cocktail components
  • Supplied as a 100X concentrate—effective when used at a final concentration of 1X; however, samples that contain particularly high levels of phosphatase activity may require a final concentration of 2-3X
  • Compatibility— compatible with most detergent-based cell lysis reagents, including all Thermo Scientific Pierce Cell Lysis Solutions
  • All-in-one format—inhibits both serine-threonine phosphatases and protein tyrosine phosphatases
  • Compatible with popular protein assays—works with standard protein assays, including BCA and coomassie (Bradford) assays
  • Convenient storage—refrigerator (2 to 8°C) storage saves valuable space in the freezer and provides for immediate use

Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from serine/threonine phosphatases as well as protein tyrosine phosphatases (PTPs) during cell lysis. The cocktail contains a mixture of four inhibitors of broad specificity, including sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. Conveniently packaged in a 100X format, the cocktail is stable at 4°C and is suitable for use with any cell or tissue extract. The cocktail preserves protein phosphorylation in immunoprecipitation and kinase assays.

Phosphorylation and dephosphorylation comprise a molecular on/off switch that regulates many key biological pathways within the cell, including signal transduction, cell division and apoptosis. To effectively study phosphorylation activation states, one must inactivate phosphatases to prevent their uncontrolled activity upon cell lysis. Halt Phosphatase Inhibitor Cocktail accomplishes this task.

Related Products

  • Pierce™ Phosphatase Inhibitor Mini Tablets (Cat. No. 88667)

Specifications

Product Type Phosphatase Inhibitor Cocktail
Content And Storage Upon receipt store product at 4°C. Do not freeze.
Form Liquid
Product Line Halt
Reagent Type Phosphatase Inhibitor
Inhibitors Phosphatase Inhibitor
Quantity 1 mL
Do you offer the components of the Halt Protease and Phosphatase Cocktail individually?

Yes. Halt Protease, Halt Phosphatase and Protease/Phosphatase combined Inhibitor Cocktails are available. In addition, some of the individual inhibitors of the Halt Protease and Phosphatase Inhibitor Cocktail are offered separately.

Is the Halt Phosphatase Inhibitor Cocktail (Cat. No. 78420) EDTA-free?

Yes, the Halt Phosphatase Inhibitor Cocktail (Cat. No. 78420) is EDTA-free. It just contains the indicated phosphatase Inhibitors.

I accidentally stored Halt Phosphatase Inhibitor Cocktail Bulk (100 mL) (Cat. No. 78420B) at -20 degrees C. Can I still use it?

We recommend storing Halt Phosphatase Inhibitor Cocktail Bulk (100 mL) at 4 degrees C. We do not recommend freezing it. One of the components of this cocktail (orthovandate) tends to form higher order structures and other oxidation states. These are colored products which can cause the cocktail to vary from colorless to yellow (and still be active). If the cocktail turns green, however, which can happen during freezing, it means that the orthovandate has lost its activity.

If the cocktail was accidentally frozen, we recommend thawing it out and examining the color. If the cocktail has turned green, we recommend discarding it.

For Research Use Only. Not for use in diagnostic procedures.