missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hafnium(IV) Trifluoromethanesulfonate 90.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| UN Number | 1759 |
| Formula Weight | 774.74 |
| Percent Purity | ≥90.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Hafnium(IV) Trifluoromethanesulfonate |
Safety and Handling
| TSCA | No |