missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gum arabic, MP Biomedicals™
Description
- It is used in textiles, ceramics, lithography, cosmetics and pharmaceuticals.
- In the food industry, acacia is mainly used as confectionery, bakery, dairy, drink, and as a microencapsulating agent.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 1 kg |
| Synonym | Arabic gum, Australian gum |
| Formula Weight | 250,000 to 1,000,000 |
| Chemical Name or Material | Gum arabic |