missing translation for 'onlineSavingsMsg'
Learn More

Guanylthiourea 98.0+%, TCI America™

Catalog No. G023425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G023425G 25 g
G0234100G 100 g
G0234500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G023425G Supplier TCI America Supplier No. G023425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2114-02-5
Molecular Formula C2H6N4S
Molecular Weight (g/mol) 118.16
MDL Number MFCD00014472
InChI Key OKGXJRGLYVRVNE-UHFFFAOYSA-N
Synonym amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine
PubChem CID 2724563
IUPAC Name diaminomethylidenethiourea
SMILES C(=NC(=S)N)(N)N

Specifications

Color White
Quantity 25 g
Formula Weight 118.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Guanylthiourea
EINECSNumber (2)-1780
RTECSNumber YR8250000
TSCA No