missing translation for 'onlineSavingsMsg'
Learn More

Guanosine, 99%

Catalog No. AC411130250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411130250 25 g Glass bottle
AC411131000 100 g Glass bottle
AC411135000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC411130250 Supplier Thermo Scientific Chemicals Supplier No. 411130250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 118-00-3 | C10H13N5O5 | 283.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-00-3
Molecular Formula C10H13N5O5
Molecular Weight (g/mol) 283.24
MDL Number MFCD00010182
InChI Key NYHBQMYGNKIUIF-UUOKFMHZSA-N
Synonym guanosine, guanine riboside, vernine, guanozin, guanosin, inosine, 2-amino, usaf cb-11, vernine van, l-guanosine, 9-beta-d-ribofuranosylguanine
PubChem CID 6802
ChEBI CHEBI:16750
SMILES NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Melting Point 250°C
Color White to Yellow
Loss on Drying 5% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Beilstein 26, V,18, 81
Merck Index 15, 4601
Specific Rotation Condition −62.00 (20.00°C c=3,in 0.1 N NaOH)
Specific Rotation -62
Solubility Information Solubility in water: slightly soluble. Other solubilities: insoluble in ethanol,ethylether,soluble in acetic acid
IUPAC Name 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Formula Weight 283.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Guanosine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 204-227-8

RUO – Research Use Only