missing translation for 'onlineSavingsMsg'
Learn More

Guanosine, 98%

Catalog No. AAA1132830
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1132830 250 g
AAA1132814 25 g
AAA1132818 50 g
AAA1132822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1132830 Supplier Thermo Scientific Chemicals Supplier No. A1132830

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 118-00-3 | C10H13N5O5 | 283.24 g/mol

Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.

Solubility
Soluble in Formic Acid:Water (1:1): 50 mg/mL. Insoluble in methanol, diethyl ether.

Notes
Store away from extreme heat. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 118-00-3
Molecular Formula C10H13N5O5
Molecular Weight (g/mol) 283.24
MDL Number MFCD00010182
InChI Key NYHBQMYGNKIUIF-UUOKFMHZSA-N
Synonym guanosine, guanine riboside, vernine, guanozin, guanosin, inosine, 2-amino, usaf cb-11, vernine van, l-guanosine, 9-beta-d-ribofuranosylguanine
PubChem CID 6802
ChEBI CHEBI:16750
SMILES NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Melting Point ∼250°C (decomposition)
Quantity 250 g
Beilstein 625911
Merck Index 14,4566
Solubility Information Soluble in Formic Acid:Water (1:1): 50mg/mL. Insoluble in methanol,diethyl ether.
Optical Rotation −72° (c=1.5 in 0.1N NaOH)
IUPAC Name 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Formula Weight 283.24
Percent Purity ≥98%
Chemical Name or Material Guanosine
Hazard Category H301
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 204-227-8
RTECSNumber MF8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only