missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Guanosine-5'-diphosphate disodium salt

Catalog No. AAJ61646ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61646ME 500 mg
AAJ61646MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61646ME Supplier Thermo Scientific Chemicals Supplier No. J61646ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7415-69-2 | C10H13N5Na2O11P2 | 487.17 g/mol

Guanosine-5'-diphosphate disodium salt is used to study the kinetics and characteristics of GTPases such as those associated with G-protein coupled receptors. It is also used to study cell signaling processes mediate by guanine nucleotide exchange factors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanosine-5′-diphosphate disodium salt is used to study the kinetics and characteristics of GTPases such as those associated with G-protein coupled receptors. It is also used to study cell signaling processes mediate by guanine nucleotide exchange factors.

Solubility
Soluble in water

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 7415-69-2
Molecular Formula C10H13N5Na2O11P2
Molecular Weight (g/mol) 487.17
MDL Number MFCD00066538
InChI Key LTZCGDIGAHOTKN-KHRSEZDTNA-L
Synonym 5-gdp pound not2na
PubChem CID 134129638
IUPAC Name disodium;(2R,3S,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolane-3,4-diolate
SMILES [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Color White
Quantity 500 mg
Solubility Information Soluble in water
Formula Weight 487.17
Physical Form Powder
Chemical Name or Material Guanosine-5'-diphosphate disodium salt
EINECSNumber 231-026-2
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only