missing translation for 'onlineSavingsMsg'
Learn More

Molecular Grade Guanidine Thiocyanate, Promega

Catalog No. PRV2791
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV2791 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV2791 Supplier Promega Corporation Supplier No. V2791

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protein denaturant used most commonly for the isolation of intact RNA due to its ability to inhibit RNase.

  • Quality tested and certified free of DNase, RNase and protease activity

Chemical Identifiers

CAS 593-84-0
Synonym Guanidinium Thiocyanate

Specifications

Color White
Quantity 100 g
Formula Weight 118.16
Grade Molecular
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Thiocyanate, For the isolation of intact RNA due to its ability to inhibit RNase
Recommended Storage 15 to 30°C