missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Thiocyanate 98.0+%, TCI America™

Catalog No. G03605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03605G 5 g
G036025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G03605G Supplier TCI America Supplier No. G03605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 593-84-0
Molecular Formula CH5N3
MDL Number MFCD00013027

Specifications

Melting Point 120°C
Color White
pH 4.5 to 7.0 (6M H2O 25°C)
Quantity 5 g
Formula Weight 118.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Thiocyanate [for Biochemical Research]
RTECSNumber XL1225000
TSCA Yes