missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Guanidine Thiocyanate, ≥99%, Calbiochem™,

Catalog No. 368975500GM
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
368975500GM 500 g Glass Bottle
368975100GM 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 368975500GM Supplier MilliporeSigma™ Supplier No. 368975500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 593-84-0
Molecular Formula CH5N3 · CHNS

Specifications

Color White
Quantity 500 g
Packaging Glass Bottle
Percent Purity ≥99% (Titration)