missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine Sulfamate 90.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 500 g |
| Formula Weight | 156.16 |
| Percent Purity | ≥90.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Guanidine Sulfamate |
Safety and Handling
| TSCA | Yes |