missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, Ultra Pure, 500g, MP Biomedicals

Catalog No. ICN820539
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN820539 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN820539 Supplier MP Biomedicals Inc Supplier No. 04820539

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

50-01-1 Purity: >>99% O.D. (260 nm) of a 6 M solution: <<0.03 Ammeline: none detected Protein denaturant for molecular weight determinations

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Quantity 500 g
Grade Ultrapure
Chemical Name or Material Guanidine Hydrochloride, Ultra Pure