missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride 98.0+%, TCI America™

Catalog No. G0162500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0162500G 500 g
G016225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0162500G Supplier TCI America Supplier No. G0162500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 186°C
Color White
pH 5.56 (50g/L H2O soln. 25°C)
Quantity 500 g
Formula Weight 95.53
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Hydrochloride
RTECSNumber MF4300000
TSCA Yes