missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Guanidine Hydrochloride, Molecular biology grade, Calbiochem™,

Catalog No. 3690801KG
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3690801KG 1 kg Glass Bottle
369080100GM 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3690801KG Supplier MilliporeSigma™ Supplier No. 3690801KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym Guanidinium Chloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Color White
Quantity 1 kg
Packaging Glass Bottle
Grade Molecular Biology