missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine Hydrochloride, 99+%, MP Biomedicals
$93.56 - $1966.36
Chemical Identifiers
| CAS | 50-01-1 |
|---|---|
| Molecular Formula | CH6ClN3 |
| Molecular Weight (g/mol) | 95.53 |
| InChI Key | PJJJBBJSCAKJQF-UHFFFAOYSA-N |
| Synonym | guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride |
| PubChem CID | 5742 |
| ChEBI | CHEBI:32735 |
| IUPAC Name | guanidine;hydrochloride |
| SMILES | C(=N)(N)N.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19482625
|
MP Biomedicals Inc
0219482625 |
25 g |
Each for $93.56
|
|
|||||
|
ICN19482680
|
MP Biomedicals Inc
0219482680 |
100 g |
Each for $163.42
|
|
|||||
|
ICN19482690
|
MP Biomedicals Inc
0219482690 |
500 g |
Each for $524.39
|
|
|||||
|
ICN19482691
|
MP Biomedicals Inc
0219482691 |
1 kg |
Each for $917.46
|
|
|||||
|
ICN19482692
|
MP Biomedicals Inc
0219482692 |
3 kg |
Each for $1,966.36
|
|
|||||
Chemical Identifiers
| 50-01-1 | |
| 95.53 | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| CHEBI:32735 | |
| C(=N)(N)N.Cl |
| CH6ClN3 | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| 5742 | |
| guanidine;hydrochloride |
Specifications
| 50-01-1 | |
| CH6ClN3 | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| C(=N)(N)N.Cl | |
| 95.53 | |
| CHEBI:32735 | |
| Guanidine Hydrochloride |
| 99+% | |
| 25 g | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| guanidine;hydrochloride | |
| 5742 | |
| 95.53 |
Safety and Handling
Recommended Storage : Room Temperature