missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine Hydrochloride, 99+%, MP Biomedicals
Supplier: MP Biomedicals Inc 0219482690
Specifications
| Guanidine Hydrochloride | |
| 99+% | |
| 500 g | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| guanidine;hydrochloride | |
| 5742 | |
| 95.53 |
| 50-01-1 | |
| CH6ClN3 | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| C(=N)(N)N.Cl | |
| 95.53 | |
| CHEBI:32735 |
Chemical Identifiers
| 50-01-1 | |
| 95.53 | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| CHEBI:32735 | |
| C(=N)(N)N.Cl |
| CH6ClN3 | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| 5742 | |
| guanidine;hydrochloride |
Safety and Handling
Recommended Storage : Room Temperature