missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, 99+%, MP Biomedicals

Supplier:  MP Biomedicals Inc 0219482691

 View more versions of this product

Catalog No. ICN19482691


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Guanidine Hydrochloride
99+%
1 kg
PJJJBBJSCAKJQF-UHFFFAOYSA-N
guanidine;hydrochloride
5742
95.53
50-01-1
CH6ClN3
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
C(=N)(N)N.Cl
95.53
CHEBI:32735

Chemical Identifiers

50-01-1
95.53
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
CHEBI:32735
C(=N)(N)N.Cl
CH6ClN3
PJJJBBJSCAKJQF-UHFFFAOYSA-N
5742
guanidine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions