missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, 99+%, MP Biomedicals

Catalog No. ICN19482692
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
3 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19482692 3 kg
ICN19482625 25 g
ICN19482680 100 g
ICN19482690 500 g
ICN19482691 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN19482692 Supplier MP Biomedicals Inc Supplier No. 0219482692

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Assay Percent Range 99+%
Quantity 3 kg
Formula Weight 95.53
Chemical Name or Material Guanidine Hydrochloride
Recommended Storage Room Temperature