missing translation for 'onlineSavingsMsg'
Learn More

Medisca Guaifenesin, USP

Numéro de catalogue. 14185662
Modifier l'affichage
Click to view available options
Quantity:
100 g
500 g
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
14185662 25 kg
14185628 500 g
14185661 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 14185662 Fournisseur Medisca Code fournisseur 069607

Guaifenesin USP is a white to slightly gray, crystalline powder. May have a slight characteristic odor. Alternative name(s): GLycerol guaiacolate, guaiphenesin

Guaifenesin USP is a white to slightly gray, crystalline powder. May have a slight characteristic odor.

Alternative name(s): GLycerol guaiacolate, guaiphenesin

Spécifications

Chemical Name or Material Guaifenesin, USP
Packaging Lid, White, PP, Rectangular, w/EZ Tear Tab (for MWP90013GAL)
CAS 93-14-1
Type Drug Standard
Molecular Weight (g/mol) 198.2166
Quantity 25 kg
Percent Purity 98 to 102 %
Grade USP